Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

S0695

Sigma-Aldrich

Anti-Somatostatin Receptor Type 2 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-SSTR2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

availability

not available in Japan

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 24 μg/mL using human pancreas, Islets of Langerhans (Optimal dilution should be determined by the researcher.)

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... SSTR2(6752)
mouse ... Sstr2(20606)
rat ... Sstr2(54305)

General description

Somatostatin receptor 2 (SSTR2) is encoded by the gene mapped to human chromosome 17q25.1. It is mainly localized on choroid plexus and arachnoid granulations. SSTR2 is transcribed into two isoforms, SSTR2A and SSTR2B through alternative splicing.

Immunogen

synthetic peptide corresponding to the second extracellular loop of human somatostatin receptor type 2, conjugated to KLH. The immunizing peptide has 100% homology with the rat and mouse gene.

Biochem/physiol Actions

Somatostatin receptor 2 (SSTR2) mediates human anterior pituitary hormone suppression by interacting with its peptide ligand somatostatin. It also plays a major role in maintenance of brain fluid homeostasis. The encoded protein has an ability to hinder the cell proliferation of solid tumors. Experimental studies states that SSTR2 methylation is presumed to be involved in the metastasis and invasion of laryngeal cancer.

Physical form

Solution in phosphate buffered saline, pH 7.7, containing 0.1% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Genetic analysis of inherited hydrocephalus in a rat model
Jones HC, et al.
Experimental Neurology, 190, 79-90 (2004)
SSTR2 promoter hypermethylation is associated with the risk and progression of laryngeal squamous cell carcinoma in males
Shen Z, et al.
Diagnostic Pathology (2016)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service