Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

S0694

Sigma-Aldrich

Anti-Somatostatin Receptor Type 2 antibody produced in rabbit

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-SS2R, Anti-SSTR2a

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

rat

technique(s)

western blot: 1:200 using rat brain membranes

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... SSTR2(6752)
mouse ... Sstr2(20606)
rat ... Sstr2(54305)

General description

Somatostatin (SST) is a peptide. Somatostatin receptor 2 (SSTR2) codes for somatostatin receptor and is abundantly expressed in normal tissues.

Immunogen

peptide corresponding to amino acid residues 339-359 of rat somatostatin receptor type 2. This epitope is identical in human, mouse, bovine, and pig.

Application

Anti-Somatostatin Receptor Type 2 antibody has been used in immunohistochemical studies.

Biochem/physiol Actions

Somatostatins performs several functions like anti-proliferation, pro-apoptosis and anti-migration. Somatostatin receptor 2 (SSTR2) can block the cell multiplication of solid tumors.

Physical form

Lyophilized at ~0.7 mg/ml in phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 0.05% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An Immunohistochemical Study on the Endocrine Cells in the Stomach and Intestine Regions of the Dicentrarchus labrax, L.,1758
Dilek D?LER
F?rat University Veterinary Journal of Health Sciences (2011)
SSTR2 promoter hypermethylation is associated with the risk and progression of laryngeal squamous cell carcinoma in males
Shen Z, et al.
Diagnostic Pathology, 11 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service