Key Documents
S0685
Silicone isolators, Press-to-Seal
1 well, D × L × W 0.8 mm × 32 mm × 19 mm, adhesive/adhesive, oblong
About This Item
UNSPSC Code:
41100000
NACRES:
MA.01
material
adhesive/adhesive
oblong
manufacturer/tradename
Grace Bio-Labs 666503
D × L × W
0.8 mm × 32 mm × 19 mm
Application
Isolate specimens on a microscope slide with hydrophobic press-to-seal (removable) or peel-and-stick (adhesive) barriers. Available in silicone thickness 0.5 to 2.5mm as pre-cut gaskets and sheet material; with or without secure seal clean release adhesive on one or both surfaces. Non-adhesive isolators are autoclavable and re-usable for cell culture applications. NOTE: Add approximately 0.15mm to depth per layer of adhesive.
Choose from one of the most recent versions:
Certificates of Analysis (COA)