Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

S0645

Sigma-Aldrich

Anti-Sodium Channel PN1 antibody produced in rabbit

affinity isolated antibody, lyophilized powder

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

rat

technique(s)

western blot (chemiluminescent): 1:200-1:400

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... SCN9A(6335)
rat ... Scn9a(78956)

General description

Sodium channel protein type 9 subunit alpha (SCN9A) is encoded in human chromosome location 2q24.3. SCN9A is also called the Nav1.7. Sodium channel protein type 9 subunit alpha, (SCN9A) encodes nine isoforms in human and they are majorly expressed in cells of peripheral nervous system.

Immunogen

synthetic peptide corresponding to amino acids 446-460 of rat PN1 (with additional N-terminal cysteine). This epitope is highly conserved in rabbit and human.

Biochem/physiol Actions

They function to generate and conduct electrical impulses in neuronal membranes. SCNA9 mutations is associated with epilepsy in infant dravet syndrome. Nonsense mutations in SCNA9 gene leads to loss sensation towards pain. Missense mutations in SCNA9 results in primary erythermalgia and paroxysmal extreme pain disorder.

Physical form

Lyophilized from a solution containing phosphate buffered saline, pH 7.4, 1% bovine serum albumin, and 0.05% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

From ionic currents to molecular mechanisms: the structure and function of voltage-gated sodium channels
Catterall WA
Neuron, 26(1), 13-25 (2000)
p. L1612P, a novel voltage-gated sodium channel Nav1. 7 mutation inducing a cold sensitive paroxysmal extreme pain disorder
Suter MR, et al.
Anesthesiology, 122(2), 414-423 (2015)
Structural basis of Nav1. 7 inhibition by an isoform-selective small-molecule antagonist
Ahuja S, et al.
Science, 350(6267), aac5464-aac5464 (2015)
International union of pharmacology. xlvii. Nomenclature and structure-function relationships of voltage-gated sodium channels
Catterall WA, et al.
Pharmacological Reviews, 57(6267), 397-409 (2005)
Epilepsy phenotype associated with a chromosome 2q24. 3 deletion involving SCN1A: Migrating partial seizures of infancy or atypical Dravet syndrome
Lim B, et al.
Epilepsy Research, 109, 34-39 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service