Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

S0451

Sigma-Aldrich

Select-HA Hyaluronan

50K, mol wt 25-75 kDa

Synonym(s):

Hyaluronic acid

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352201

Quality Level

mol wt

25-75 kDa

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Select-HA is hyaluronic acid (HA) made through enzymatic synthesis where a very high level of size control is possible. Each Select-HA is a narrow-size distribution HA with a specific average molecular weight within a defined range. Polydispersity of Select-HA is as low as 1.02 and averages 1.1 (a value of 1 is for a ‘perfect′ polymer). In contrast, the polydispersity of most other commercial HA is usually greater than 2. Protein content is <0.1% for all Select-HA products.

Analysis Note

The molecular mass of each Select-HA is determined by multi-angle laser light scattering-size exclusion chromatography (MALLS-SEC). Actual result is reported on the lot-specific certificate of analysis. Sold on the basis of mg hyaluronan as determined by carbazole assay.

Legal Information

Select-HA is a trademark of Hyalose LLC

Storage Class

11 - Combustible Solids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service