Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

RN0100

Sigma-Aldrich

Ribonuclease A Activity Kit

sufficient for 100 tests

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.26

usage

sufficient for 100 tests

storage temp.

2-8°C

General description

Ribonuclease A (RNase A), a well-known protein, is frequently used as a model system to study folding, stability, and chemistry.[1] It is made up of three α helices and seven β strands that are organized in two "lobes".[2]

Application

Ribonuclease A Activity Kit has been used to assess the activity of RNase released from the nanogels.[3]

Biochem/physiol Actions

Ribonuclease A (RNase A) can convert RNA to oligonucleotides in the presence of water. In the absence of metal ions or cofactors, RNase A can catalyze the hydrolysis of single-stranded RNA.[4]

Kit Components Also Available Separately

Product No.
Description
SDS

  • Ribonuclease A Reaction Buffer

  • R5500Ribonuclease A from bovine pancreas, Type XII-A, ≥90% (SDS-PAGE), 75-125 Kunitz units/mg proteinSDS

  • R6750Ribonucleic acid from baker’s yeast (S. cerevisiae)SDS

pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Resp. Sens. 1

Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lifelike but Not Living: Selection of Synthetically Modified Bioinspired Nucleic Acids for Binding and Catalysis
Perrin, D. M.
Polymer Science: A Comprehensive Reference (2012)
Xinghui Si et al.
Journal of controlled release : official journal of the Controlled Release Society, 320, 83-95 (2020-01-20)
As the most common malignancy in women, breast cancer causes >40,000 deaths annually. Ribonuclease A (RNase), a new anti-cancer agent, has attracted intense interest due to its high efficacy and specificity. However, RNase suffers from instability, a short half-life in
Predicting pKa Values with Continuous Constant pH Molecular Dynamics
Wallace JA and Shen JK
Methods in Enzymology (2009)
Bovine Pancreatic Ribonuclease A: Oxidative and Conformational Folding Studies
Scheraga HA, et al.
Methods in Enzymology (2001)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service