Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

RES1520C-A7

SAFC

L-Cystine

Synonym(s):

L-Cystine, (R,R)-3,3′-Dithiobis(2-aminopropionic acid)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-SCH2CH(NH2)CO2H]2
CAS Number:
Molecular Weight:
240.30
Beilstein/REAXYS Number:
1728094
EC Number:
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.21

biological source

plant

assay

98.5-101.0% dry basis

form

powder

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

mp

>240 °C (dec.) (lit.)

suitability

suitable for manufacturing use

foreign activity

Cytotoxicity; tested

SMILES string

N[C@@H](CSSC[C@H](N)C(O)=O)C(O)=O

InChI

1S/C6H12N2O4S2/c7-3(5(9)10)1-13-14-2-4(8)6(11)12/h3-4H,1-2,7-8H2,(H,9,10)(H,11,12)/t3-,4-/m0/s1

InChI key

LEVWYRKDKASIDU-IMJSIDKUSA-N

Looking for similar products? Visit Product Comparison Guide

General description

To request documentation for this product, please contact Customer Support and select ‘Product Documentation′. Please note that access to documentation for this product requires a confidentiality disclosure agreement.

Application

L-Cystine is a sulfur containing non-essential amino acid. It is used as a cell culture media component and raw material for the commercial biomanufacture of therapeutic recombinant proteins and monoclonal antibodies.

Packaging

Product is available in the following package sizes:
RES1520C-A701X: 100 gm container
RES1520C-A702X: 1 kg container
RES1520C-A704X: 10 kg container
RES1520C-A705X: 25 kg container

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Richard Bridges et al.
Pharmacological reviews, 64(3), 780-802 (2012-07-05)
System x(c)(-) represents an intriguing target in attempts to understand the pathological states of the central nervous system. Also called a cystine-glutamate antiporter, system x(c)(-) typically functions by exchanging one molecule of extracellular cystine for one molecule of intracellular glutamate.
Masa Cemazar et al.
Antioxidants & redox signaling, 10(1), 103-111 (2007-10-27)
Cyclic cystine knot proteins are small but topologically complex molecules that occur naturally in plants and have a wide range of bioactivities that make them interesting from a pharmaceutical perspective. Their remarkable stability is dependent on the correct formation of
Maisie Lo et al.
Journal of cellular physiology, 215(3), 593-602 (2008-01-09)
The x(c) (-) cystine/glutamate antiporter is a major plasma membrane transporter for the cellular uptake of cystine in exchange for intracellular glutamate. Its main functions in the body are mediation of cellular cystine uptake for synthesis of glutathione essential for
Jan Lewerenz et al.
Antioxidants & redox signaling, 18(5), 522-555 (2012-06-07)
The antiporter system x(c)(-) imports the amino acid cystine, the oxidized form of cysteine, into cells with a 1:1 counter-transport of glutamate. It is composed of a light chain, xCT, and a heavy chain, 4F2 heavy chain (4F2hc), and, thus
Serge-George Rosolen et al.
Advances in experimental medicine and biology, 775, 145-154 (2013-02-09)
In the 70s, the amino acid taurine was found essential for photoreceptor survival. Recently, we found that taurine depletion can also trigger retinal ganglion cell degeneration both in vitro and in vivo. Therefore, evaluation of taurine levels could be a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service