Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

RAB0813

Sigma-Aldrich

Mouse Il23a / Interleukin-23 Subunit Alpha ELISA Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

species reactivity

mouse

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type serum
sample type plasma
sample type cell culture supernatant(s)

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 25 pg/mL
standard curve range: 24.58-6000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... Il23a(83430)

General description

The antibody pair provided in this kit recognizes Mouse interleukin 23 alpha subunit.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

pictograms

Corrosion

signalword

Warning

hcodes

Hazard Classifications

Met. Corr. 1

Storage Class

8A - Combustible corrosive hazardous materials

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Dionne et al.
Mucosal immunology, 7(6), 1405-1415 (2014-05-02)
Accumulating evidence implicates defective innate immunity in the pathogenesis of Crohn's disease (CD). Ineffectual NOD2 (nucleotide-binding oligomerization domain 2) is the most common susceptibility gene contributing to CD. Vitamin D (vD), a potent modulator of innate and adaptive immunity, induces
Heng-Xiu Yan et al.
Immunologic research, 60(1), 112-126 (2014-06-05)
Psoriasis is a common chronic T-cell-mediated autoimmune skin disease, and traditional immunotherapies for psoriasis have focused on the direct inhibition of T cells, which often causes toxicity and lacks long-term effectiveness. Safe and effective therapeutic strategies are strongly needed for
Anna Olewicz-Gawlik et al.
International journal of rheumatic diseases, 17(6), 664-670 (2014-01-29)
T cell abnormalities with a focus on Th17 cells have been associated with the pathogenesis of systemic sclerosis (SSc) and interstitial lung disease (ILD). The aim of this study was to evaluate serum levels of interleukin (IL)-17, IL-21 and IL-23
Kobporn Boonnak et al.
Journal of immunology (Baltimore, Md. : 1950), 192(12), 5906-5912 (2014-05-16)
Although lymphopenia is a hallmark of severe infection with highly pathogenic H5N1 and the newly emerged H7N9 influenza viruses in humans, the mechanism(s) by which lethal H5N1 viruses cause lymphopenia in mammalian hosts remains poorly understood. Because influenza-specific T cell
Indumathi Manoharan et al.
Journal of immunology (Baltimore, Md. : 1950), 193(8), 4203-4213 (2014-09-12)
Dendritic cells (DCs) sense microbes via multiple innate receptors. Signals from different innate receptors are coordinated and integrated by DCs to generate specific innate and adaptive immune responses against pathogens. Previously, we have shown that two pathogen recognition receptors, TLR2

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service