Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

RAB0730

Sigma-Aldrich

Human S100A8 / Protein S100-A8 ELISA Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type serum
sample type cell culture supernatant(s)
sample type plasma

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 10 pg/mL
standard curve range: 10.24-2500 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... S100A8(6279)

General description

The S100A8 ELISA kit provides for the Quantitative measurement of S100A8 in Cell Culture Supernatants, Plasma and Serum.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

pictograms

Corrosion

signalword

Warning

hcodes

Hazard Classifications

Met. Corr. 1

Storage Class

8A - Combustible corrosive hazardous materials

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stanley Moore et al.
The Journal of rheumatology, 47(11), 1652-1660 (2019-12-17)
Neutrophil extracellular traps (NET) are essential in host defense, but are also linked to inflammation and autoimmunity, including in systemic lupus erythematosus (SLE). We recently described that immune complexes (IC) induce NET formation, promoting SLE-like disease in mice. In the
Valentina Giudice et al.
Cytokine, 113, 462-465 (2018-07-01)
The alarmin family members S100A8 and S100A9 are acute phase inflammation proteins, but they also have been proposed as biomarkers in many malignant and non-malignant diseases. In this study, circulating S100A8 and S100A9 homodimers and S100A8/A9 heterodimers in plasma were
Shruti Bhattacharya et al.
Pediatric rheumatology online journal, 18(1), 6-6 (2020-01-17)
Axial SpA and Enthesitis related arthritis (ERA) patients show strong HLA-B27 association, gut dysbiosis, high toll like receptor (TLR)2 and 4 expression on monocytes, pro-inflammatory cytokine production and elevated levels of TLR4 endogenous ligands [tenascin-c (TNC) and myeloid related protein
Tim Erkens et al.
Journal of applied toxicology : JAT, 38(10), 1282-1292 (2018-05-22)
The intestinal barrier controls intestinal permeability, and its disruption has been associated with multiple diseases. Therefore, preclinical safety biomarkers monitoring barrier integrity are essential during the development of drugs targeting the intestines, particularly if starting treatment early after onset of
Yu-Qin Wang et al.
Ocular immunology and inflammation, 28(1), 133-141 (2018-11-06)
Purpose: To investigate whether there is an association between circulating S100A8/A9 levels and uveitis activity.Methods: A total of 549 plasma samples were collected from uveitis patients and non-uveitic controls.Results: S100A8/A9 plasma levels were elevated in uveitis patients compared to non-uveitic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service