Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

RAB0662

Sigma-Aldrich

Human GAS1 / Growth Arrest-specific Protein 1 ELISA Kit

Sign Into View Organizational & Contract Pricing

Select a Size

96 WELLS
$569.00

$569.00


Usually Ships in 4 Weeks. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
96 WELLS
$569.00

About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

$569.00


Usually Ships in 4 Weeks. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type serum
sample type cell culture supernatant(s)
sample type plasma

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 45 pg/mL
standard curve range: 46.88-3000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... GAS1(2619)

General description

The antibody pair provided in this kit recognizes human Growth Arrest-specific Protein 1.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

pictograms

Corrosion

signalword

Warning

hcodes

Hazard Classifications

Met. Corr. 1

Storage Class

8A - Combustible corrosive hazardous materials

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laura Sánchez-Hernández et al.
Gene therapy, 25(6), 439-449 (2018-06-27)
The overexpression of GAS1 (Growth Arrest Specific 1) in glioma cells induces cell cycle arrest and apoptosis. We previously demonstrated that the apoptotic process set off by GAS1 is caused by its capacity to inhibit the Glial cell-derived neurotrophic factor
Lizbeth Daniel-García et al.
OncoTargets and therapy, 13, 11769-11779 (2020-11-26)
Pancreatic carcinoma cells exhibit a pronounced tendency to invade along and through intra and extrapancreatic nerves, even during the early stages of the disease, a phenomenon called perineural invasion (PNI). Thus, we sought to determine the effects of the simultaneous

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service