Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

RAB0596

Sigma-Aldrich

Feline PDGFB / Platelet-derived Growth Factor Subunit B ELISA Kit

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
$655.00

$655.00


Usually Ships in 4 Weeks. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
1 KIT
$655.00

About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

$655.00


Usually Ships in 4 Weeks. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

species reactivity

feline

technique(s)

ELISA: suitable

assay range

inter-assay cv: <10%
intra-assay cv: <12%

shipped in

wet ice

storage temp.

−20°C

General description

The antibody pair provided in this kit recognizes Feline Platelet-derived growth factor subunit B.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

pictograms

Corrosion

signalword

Warning

hcodes

Hazard Classifications

Met. Corr. 1

Storage Class

8A - Combustible corrosive hazardous materials

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service