Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

RAB0419

Sigma-Aldrich

Human Resistin ELISA Kit

for serum, plasma, cell culture supernatant and urine

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
$569.00

$569.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
1 KIT
$569.00

About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

$569.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type plasma
sample type urine
sample type cell culture supernatant(s)
sample type serum

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 2 pg/mL
standard curve range: 1.4-400 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... RETN(56729)

General description

The Human Resistin ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human Resistin in serum, plasma, cell culture supernatants and urine.

Immunogen

Recombinant Human Resistin

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABELADAELISA 1X Assay/Sample Diluent Buffer A (Item D1)SDS

  • RABELADBELISA 5X Assay/Sample Diluent Buffer B (Item E1)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

pictograms

Corrosion

signalword

Warning

hcodes

Hazard Classifications

Met. Corr. 1

Storage Class

8A - Combustible corrosive hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vanessa S Fontes et al.
Revista da Associacao Medica Brasileira (1992), 66(3), 300-306 (2020-06-11)
To compare the serum concentrations of adipokines resistin and chemerin in children and adolescents with eutrophic and overweight and to evaluate their relationship with anthropometric, biochemical, and blood pressure variables. a cross-sectional epidemiological study was conducted with 234 students enrolled
Worrawalan Phoonsawat et al.
Biochemical and biophysical research communications, 448(3), 261-266 (2014-05-06)
Exosomes are membrane vesicles 30-120 nm in diameter that are released by many cell types and carry a cargo of proteins, lipids, mRNA, and microRNA. Cultured adipocytes reportedly release exosomes that may play a role in cell-to-cell communication during the
Shalini Behl et al.
Molecular biology reports, 46(3), 2643-2655 (2019-03-31)
Rilpivirine is a non-nucleoside reverse transcriptase inhibitor, recently developed as a drug of choice for initial anti-retroviral (ARV) treatment of HIV-1 infection, whereas estradiol is a major component of hormonal contraceptives. Both drugs have effects on lipid metabolism, impairment of
Susann Blüher et al.
Obesity (Silver Spring, Md.), 22(7), 1701-1708 (2014-03-20)
Exercise improves weight status and metabolism. Irisin, a novel myokine, may be involved in the regulation of metabolic function. The effect of an exercise and dietary lifestyle intervention for 1-year on irisin, adipokines (leptin, adiponectin, resistin) and inflammatory markers (C-reactive
Georgia Kassi et al.
Immunological investigations, 43(6), 544-555 (2014-03-26)
Zoledronic acid is known to induce a transient acute phase response (APR). The aim of the study was to investigate whether an APR caused by zoledronic acid administration can induce insulin resistance in post-menopausal osteoporotic women and the potential involvement

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service