RAB0320
Human IL-8 / CXCL8 ELISA Kit
for cell and tissue lysates
Synonym(s):
C-X-C motif chemokine 8, Granulocyte chemotactic protein 1, IL-8, Interleukin-8, Monocyte-derived neutrophil chemotactic factor, Monocyte-derived neutrophil-activating peptide, Neutrophil-activating protein 1, T-cell chemotactic factor
Select a Size
$569.00
Select a Size
About This Item
$569.00
Recommended Products
species reactivity
human
packaging
kit of 96 wells (12 strips x 8 wells)
technique(s)
ELISA: suitable
capture ELISA: suitable
input
sample type cell lysate
sample type tissue lysate
assay range
inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 1 pg/mL
standard curve range: 0.8-600 pg/mL
detection method
colorimetric
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... IL8(3576)
General description
Immunogen
application
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Other Notes
Please type the word sample in the text box provided for lot number.
Kit Components Also Available Separately
recommended
signalword
Warning
hcodes
pcodes
Hazard Classifications
Met. Corr. 1
Storage Class
8A - Combustible corrosive hazardous materials
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service