Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

RAB0313

Sigma-Aldrich

Human IL-6 sR ELISA Kit

for serum, plasma, cell culture supernatant and urine

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
$569.00

$569.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
1 KIT
$569.00

About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

$569.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type urine
sample type serum
sample type cell culture supernatant(s)
sample type plasma

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 3 pg/mL
standard curve range: 4.1-1000 pg/mL

detection method

colorimetric

shipped in

wet ice

Storage temp.

−20°C

Gene Information

human ... IL6R(3570)

General description

The Human IL-6 sR (interleukin 6 soluble receptor) ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human IL-6 sR in serum, plasma, cell culture supernatants and urine.

Immunogen

Recombinant Human IL6R

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABELADAELISA 1X Assay/Sample Diluent Buffer A (Item D1)SDS

  • RABELADBELISA 5X Assay/Sample Diluent Buffer B (Item E1)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

pictograms

Corrosion

signalword

Warning

hcodes

Hazard Classifications

Met. Corr. 1

Storage Class

8A - Combustible corrosive hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Xaquin Castro Dopico et al.
Nature communications, 6, 7000-7000 (2015-05-13)
Seasonal variations are rarely considered a contributing component to human tissue function or health, although many diseases and physiological process display annual periodicities. Here we find more than 4,000 protein-coding mRNAs in white blood cells and adipose tissue to have
Jia Shen et al.
Experimental and therapeutic medicine, 24(3), 564-564 (2022-08-19)
The aim of the present study was to investigate the effects of microRNA (miR)-374a-5p on sepsis-induced acute lung injury (ALI) and the associated mechanism. Lipopolysaccharide (LPS)-induced human pulmonary microvascular endothelial cells (HPMVECs) were used to construct the cellular model of
Jean Gaschignard et al.
Clinical infectious diseases : an official publication of the Infectious Diseases Society of America, 59(2), 244-251 (2014-04-25)
About 10% of pediatric patients with invasive pneumococcal disease (IPD) die from the disease. Some primary immunodeficiencies (PIDs) are known to confer predisposition to IPD. However, a systematic search for these PIDs has never been carried out in children presenting
K Hultman et al.
Journal of thrombosis and haemostasis : JTH, 12(5), 701-712 (2014-03-13)
Excess fibrin in blood vessels is cleared by plasmin, the key proteolytic enzyme in fibrinolysis. Neurological disorders and head trauma can result in the disruption of the neurovasculature and the entry of fibrin and other blood components into the brain
Christina Purnama et al.
European journal of immunology, 44(7), 2003-2012 (2014-04-02)
Alveolar macrophages (AMs), localized at the pulmonary air-tissue interface, are one of the first lines of defense that interact with inhaled airborne pathogens such as influenza viruses. By using a new CD169-DTR transgenic mouse strain we demonstrate that specific and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service