Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

RAB0201

Sigma-Aldrich

GLP-1 EIA Kit

for serum, plasma, culture supernatant and cell lysates

Synonym(s):

Glucagon-like-peptide 1

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
$540.00

$540.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
1 KIT
$540.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

$540.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

species reactivity

human, rat, mouse

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
enzyme immunoassay: suitable

input

sample type cell lysate
sample type plasma
sample type serum
sample type culture supernatant(s)

assay range

inter-assay cv: <15%
intra-assay cv: <10%
sensitivity: 1.17 pg/mL
standard curve range: 0.1–1000 pg/mL

detection method

colorimetric

shipped in

wet ice

Storage temp.

−20°C

Gene Information

human ... GCG(2641)

General description

The GLP-1 Enzyme Immunoassay (EIA) Kit is an in vitro quantitative assay for detecting GLP-1 peptide based on the principle of Competitive Enzyme Immunoassay.

Immunogen

Glucagon-like Peptide 1 (GLP-1), synthetic peptide

Application

For research use only. Not for use in diagnostic procedures.

Biochem/physiol Actions

Glucagon-Like Peptide I (GLP1) controls the metabolism of glucose by inducing the secretion of insulin and suppressing that of glucagon. Studies in Indonesian population show that reduced levels of this protein are linked with increased susceptibility to type 2 diabetes mellitus.[1] Plasma levels of this hormone also show significant association with systolic and diastolic blood pressure levels.[2]

Kit Components Also Available Separately

Product No.
Description
SDS

Pictograms

Corrosion

signalword

Warning

hcodes

Hazard Classifications

Met. Corr. 1

Storage Class

8A - Combustible corrosive hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ai-Jun Li et al.
American journal of physiology. Regulatory, integrative and comparative physiology, 310(8), R724-R732 (2016-01-23)
Mercaptoacetate (MA) is an orexigenic agent reported to block fatty acid (FA) oxidation. Recently, however, we reported evidence from isolated nodose ganglion neurons that MA antagonizes the G protein-coupled long- and medium-chain FA receptor GPR40. GPR40 mediates FA-induced secretion of
Enteroendocrine cells switch hormone expression along the crypt-to-villus BMP signalling gradient
Beumer J, et al.
Nature Cell Biology, 20(8), 909-909 (2018)
Maria F Fernandes et al.
International journal of molecular sciences, 23(8) (2022-04-24)
Glucagon-like peptide-1 (GLP-1) potentiates glucose-stimulated insulin secretion (GSIS). While dozens of compounds stimulate GLP-1 secretion, few inhibit. Reduced GLP-1 secretion and impaired GSIS occur in chronic inflammation. Lysophosphatidic acids (LPAs) are bioactive phospholipids elevated in inflammation. The aim of this
Mercaptoacetate blocks fatty acid-induced GLP-1 secretion in male rats by directly antagonizing GPR40 fatty acid receptors
Li AJ, et al.
American Journal of Physiology. Regulatory, Integrative and Comparative Physiology, 310(8), R724-R732 (2016)
Jennifer O Adeghate et al.
Cells, 10(8) (2021-08-28)
The effects of early (5-day) onset of diabetes mellitus (DM) on retina ultrastructure and cellular bioenergetics were examined. The retinas of streptozotocin-induced diabetic rats were compared to those of non-diabetic rats using light and transmission electron microscopy. Tissue localization of

Questions

1–2 of 2 Questions  
  1. Which forms of GLP-1 are measured by the GLP ELISA (RAB0201)? Is it specific to the active form 7-36aa?

    1 answer
    1. The kit RAB0201 is specific for amino acids 108-128 (P01275). This includes the active forms 7-36a and 7-37, as well as the inactive cleaved form 9-36a.

      Helpful?

  2. Hi, 1 unit GLP-1 EIA Kit can be used for how many sample of blood?

    1 answer
    1. The GLP-1 EIA Kit includes a 96-well strip plate. The assay requires the addition of a blank, six standard curve points and a positive control. It is recommend to run all standards, controls and samples in duplicate. After the addition of the blank, standards and controls, each assay can run 44 samples in duplicate.

      It is recommended to perform a 2-fold sample dilution for serum/plasma samples. The sample volume required per well is 100 ul of 1:2 diluted samples. Therefore 62.5 ul of starting sample volume is sufficient to run each sample in duplicate.

      For more information on the product , please view the product data sheet: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/159/844/rab0201bul.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service