Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

RAB0054

Sigma-Aldrich

Human MCP-1 / CCL2 ELISA Kit

for serum, plasma, cell culture supernatant and urine

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
$569.00

$569.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
1 KIT
$569.00

About This Item

UNSPSC Code:
41116158

$569.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type serum
sample type cell culture supernatant(s)
sample type plasma

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... CCL2(6347)

General description

The Human MCP-1 ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human MCP-1 in serum, plasma, cell culture supernatants and urine.

Immunogen

Recombinant Human MCP1

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABELADEELISA 5X Assay/Sample Diluent Buffer E (Item E2)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

pictograms

Corrosion

signalword

Warning

hcodes

Hazard Classifications

Met. Corr. 1

Storage Class

8B - Non-combustible corrosive hazardous materials

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

JiaYi Xu et al.
Medical science monitor : international medical journal of experimental and clinical research, 26, e919854-e919854 (2020-02-07)
BACKGROUND Hypertension could induce many serious diseases, including damage to vascular endothelial cells. As a non-coding RNA, long noncoding RNA (lncRNA) has received much attention in scientific research and has a regulating efficacy on many critical life activities in human
Ka-Kyung Kim et al.
Journal of materials chemistry. B, 8(5), 945-950 (2020-01-11)
Cytokine-directed monocyte infiltration is involved in multiple pathological processes. Immuno-isolating matrices that can sequester cell-released chemokines in a microenvironment may prolong the viability and functionality of implanted materials. We describe a self-assembling peptide-based hydrogel that can capture the cytokine CCL2
Kristina Schaedlich et al.
Scientific reports, 11(1), 7928-7928 (2021-04-14)
In the member countries of the Organization for Economic Co-operation and Development (OECD), overweight and obesity affect the majority of the population. The use of environmental chemicals, such as the plasticizer DEHP, has largely increased simultaneously with this development. DEHP
Ruozhi Zhao et al.
The Journal of nutritional biochemistry, 25(8), 851-857 (2014-06-14)
Atherosclerotic cardiovascular complications are the leading cause of death in diabetic patients. Monocyte adhesion is an early event for atherogenesis. Previous studies demonstrated that dark-skin berries had cardiovascular protective effects. We hypothesize that Saskatoon berry (SB) powder may reduce monocyte
Zhenyu Yao et al.
Journal of biomedical materials research. Part A, 102(9), 3291-3297 (2013-10-15)
Wear particles generated from total joint replacements can stimulate macrophages to release chemokines, such as monocyte chemoattractant protein 1 (MCP-1), which is the most important chemokine regulating systemic and local cell trafficking and infiltration of monocyte/macrophages in chronic inflammation. One

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service