RA481500-EP
Rotor for Eppendorf® 5804/5804R & 5810/5810R Centrifuges
A-4-81, for 5810/5810R, aerosol-tight, with 4 × 500 mL rectangular buckets
Select a Size
$4,420.00
Select a Size
About This Item
$4,420.00
Recommended Products
feature
to hold 0.2 to 500 mL tubes and bottles
aerosol-tight
for 5810/5810R
medium to high-capacity swing-bucket rotor
with 4 × 500 mL rectangular buckets
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 022638602 (NA)
Eppendorf® 5810718007 (EU)
parameter
3,220 × g max. RCF
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models 5804/5804R/5810/5810R
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)