Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

R9901

Sigma-Aldrich

Tissue slide

from rat kidney, single organ

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rat kidney

Looking for similar products? Visit Product Comparison Guide

Application

Suitable for immunohistochemistry and in situ hybridization. May be used for cellular localization of tissue specific mRNA and protein expression.

Other Notes

Each slide contains a single tissue.

Preparation Note

Paraffin embeded tissue sections, approximately 5 μm thick.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service