Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

R9650

Sigma-Aldrich

Anti-RICK, C-Terminal antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-CARDIAK, Anti-RIP2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~50 kDa

species reactivity

mouse, human

concentration

~1.0 mg/mL

technique(s)

ELISA: suitable
immunocytochemistry: suitable
immunofluorescence: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RIPK2(8767)
mouse ... Ripk2(192656)
rat ... Ripk2(362491)

Immunogen

synthetic peptide amino acids 508-522 of C-terminal of human RIP-like interacting CLARP kinase (RICK).

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service