R9640
Barnstead/Thermolyne® Dewars, Locator® series
Four-wheel roller base
About This Item
Recommended Products
manufacturer/tradename
Barnstead International AY509X1
General description
Features:
- Three sizes fit the needs of almost any research lab
- Advanced vacuum insulation provides a maximum −190°C throughout flask with as little as 2 inches of liquid nitrogen, and delivers improved liquid nitrogen consumption rates
- Secure locking clasp prevents unauthorized access
- Each Locator vessel includes stainless steel racks that accommodate Nalgene Cryoboxes for 2 mL freezing vials. Cryoboxes not included
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)