Key Documents
R9384
Texan reagent reservoir for multichannel pipettes
lid: no, sterile, case of 20 x bags of 5 reservoirs
Synonym(s):
multichannel pipette reservoir, reagent reservoir
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
sterility
sterile
feature
lid: no
manufacturer/tradename
Excel Scientific RSV-0175-S05
packaging
case of 20 x bags of 5 reservoirs
capacity
175 mL
General description
Texan reagent reservoirs are reusable -- an environmentally sound alternative to disposable reagent reservoirs. Their extra-large capacity means fewer refills and time saved. The V-shaped basin is compatible with both eight- and twelve-place multiple pipettors with minimal reagent waste. Lids are removable -- attached with ball and socket hinges -- and latch tightly for secure storage of left-over reagents. With lids closed, Texan reagent reservoirs are stackable. A perimeter ridge on the lid prevents sliding. Construction is clear, chemical-resistant, autoclavable polypropylene. Molded graduation marks provide volume indication at 25, 50, 75, 100, 125, and 150 mL.
Legal Information
Texan is a trademark of Excel Scientific, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)