Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

R8906

Sigma-Aldrich

Anti-Rab10 antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-GTP-binding protein RAB10, Anti-Ras-related GTP-binding protein RAB10

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~23 kDa

species reactivity

human, rat, mouse, canine

concentration

~1.0 mg/mL

technique(s)

indirect immunofluorescence: 2-5 μg/mL using using rat NRK cells fixed and permeabilized with 4% paraformaldehyde followed by 0.4% saponin
western blot: 1-2 μg/mL using whole extract of dog MDCK cells
western blot: 2-4 μg/mL using whole extract of human A431 cells
western blot: 5-10 μg/mL using whole extract of mouse brain

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RAB10(10890)
mouse ... Rab10(19325)
rat ... Rab10(50993)

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christine Hoffmann et al.
Cellular microbiology, 16(7), 1034-1052 (2014-01-01)
The opportunistic pathogen Legionella pneumophila employs the Icm/Dot type IV secretion system and ∼300 different effector proteins to replicate in macrophages and amoebae in a distinct 'Legionella-containing vacuole' (LCV). LCVs from infected RAW 264.7 macrophages were enriched by immuno-affinity separation
Tanzina Tanu et al.
RNA biology, 18(sup1), 537-547 (2021-09-03)
Many long noncoding RNAs (lncRNAs) are localized in the nucleus and play important roles in various biological processes, including cell proliferation, differentiation and antiviral response. Yet, it remains unclear how some nuclear lncRNAs are turned over. Here we show that
Zhipeng Li et al.
Science advances, 2(12), e1601470-e1601470 (2016-12-29)
The autophagic digestion of lipid droplets (LDs) through lipophagy is an essential process by which most cells catabolize lipids as an energy source. However, the cellular machinery used for the envelopment of LDs during autophagy is poorly understood. We report
Bernhard Steiner et al.
EMBO reports, 18(10), 1817-1836 (2017-08-25)
The pathogenic bacterium Legionella pneumophila replicates in host cells within a distinct ER-associated compartment termed the Legionella-containing vacuole (LCV). How the dynamic ER network contributes to pathogen proliferation within the nascent LCV remains elusive. A proteomic analysis of purified LCVs

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service