Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

R8883

Sigma-Aldrich

Rifampicin

powder, γ-irradiated

Synonym(s):

3-(4-Methylpiperazinyliminomethyl)rifamycin SV, Rifampin, Rifamycin AMP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C43H58N4O12
CAS Number:
Molecular Weight:
822.94
Beilstein/REAXYS Number:
5723476
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Product R8883 is not currently sold in your country. Contact Technical Service

biological source

synthetic

sterility

γ-irradiated

form

powder

potency

≥880 μg per mg

color

orange to dark red

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria
mycobacteria

mode of action

protein synthesis | interferes

storage temp.

−20°C

SMILES string

CO[C@H]1\C=C\O[C@@]2(C)Oc3c(C)c(O)c4c(O)c(NC(=O)C(C)=C\C=C\[C@H](C)[C@H](O)[C@@H](C)[C@@H](O)[C@@H](C)[C@H](OC(C)=O)[C@@H]1C)c(\C=N\N5CCN(C)CC5)c(O)c4c3C2=O

InChI

1S/C43H58N4O12/c1-21-12-11-13-22(2)42(55)45-33-28(20-44-47-17-15-46(9)16-18-47)37(52)30-31(38(33)53)36(51)26(6)40-32(30)41(54)43(8,59-40)57-19-14-29(56-10)23(3)39(58-27(7)48)25(5)35(50)24(4)34(21)49/h11-14,19-21,23-25,29,34-35,39,49-53H,15-18H2,1-10H3,(H,45,55)/b12-11+,19-14+,22-13-,44-20+/t21-,23+,24+,25+,29-,34-,35+,39+,43-/m0/s1

InChI key

JQXXHWHPUNPDRT-WLSIYKJHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: macrolide

Application

Rifampicin is used as an inhibitor of transcription initiation, blocking initiation of RNA synthesis.

Biochem/physiol Actions

Inhibits the assembly of DNA and protein into mature virus particles.
Mode of Action: Inhibits initiation of RNA synthesis by binding to β-subunit of RNA polymerase.

Packaging

150mg

Preparation Note

Prepare stock solutions directly in the vial with methanol (25 mg/mL).

Other Notes

Keep container tightly closed in a dry and well-ventilated place.Product is sensitive to light and moisture. Store under inert gas. Keep in a dry place.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Lact. - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sandra V Kik et al.
The Journal of infectious diseases, 211 Suppl 2, S58-S66 (2015-03-15)
The potential available market (PAM) for new diagnostics for tuberculosis that meet the specifications of the high-priority target product profiles (TPPs) is currently unknown. We estimated the PAM in 2020 in 4 high-burden countries (South Africa, Brazil, China, and India)
Samy Figueiredo et al.
Antimicrobial agents and chemotherapy, 53(6), 2657-2659 (2009-03-25)
Two clonally related Acinetobacter baumannii isolates, A1 and A2, were obtained from the same patient. Isolate A2, selected after an imipenem-containing treatment, showed reduced susceptibility to carbapenems. This resistance pattern was related to insertion of the ISAba1 element upstream of
S Harbarth et al.
The Journal of antimicrobial chemotherapy, 70(1), 264-272 (2014-09-12)
The therapeutic arsenal for MRSA infections is limited. The aim of this study was to assess the non-inferiority of a combination of trimethoprim/sulfamethoxazole plus rifampicin versus linezolid alone for the treatment of MRSA infection. We conducted a randomized, open-label, single-centre
Anil Koul et al.
Nature chemical biology, 3(6), 323-324 (2007-05-15)
The diarylquinoline R207910 (TMC207) is a promising candidate in clinical development for the treatment of tuberculosis. Though R207910-resistant mycobacteria bear mutations in ATP synthase, the compound's precise target is not known. Here we establish by genetic, biochemical and binding assays
Rodney Dawson et al.
Lancet (London, England), 385(9979), 1738-1747 (2015-03-22)
New antituberculosis regimens are urgently needed to shorten tuberculosis treatment. Following on from favourable assessment in a 2 week study, we investigated a novel regimen for efficacy and safety in drug-susceptible and multidrug-resistant (MDR) tuberculosis during the first 8 weeks

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service