Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

R8782

Sigma-Aldrich

Ro4368554

≥98% (HPLC)

Synonym(s):

3-benzenesulfonyl-7-(4-methyl-piperazin-1-yl)-1H-indole, 7-(4-methyl-1-piperazinyl)-3-(phenylsulfonyl)-1H-Indole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H21N3O2S
CAS Number:
Molecular Weight:
355.45
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

assay

≥98% (HPLC)

form

powder

color

off-white

solubility

DMSO: >10 mg/mL

originator

Roche

Storage temp.

room temp

SMILES string

CN1CCN(CC1)c2cccc3c(c[nH]c23)S(=O)(=O)c4ccccc4

InChI

1S/C19H21N3O2S/c1-21-10-12-22(13-11-21)17-9-5-8-16-18(14-20-19(16)17)25(23,24)15-6-3-2-4-7-15/h2-9,14,20H,10-13H2,1H3

Inchi Key

AOPYPEADLGTXRA-UHFFFAOYSA-N

Biochem/physiol Actions

Ro4368554 is a 5-HT6 receptor antagonist.
Ro4368554 is a 5-HT6 receptor antagonist. RO4368554 acts selectively at 5-HT6 receptors where it binds with a greater than 100-fold selectivity over other monoamine receptor subtypes (-log M pKi = 9.4 at 5-HT6 and 7.1 at 5-HT2A).

Features and Benefits

This compound was developed by Roche. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service