R8629
Reactive Brown 10−Agarose
lyophilized powder (stabilized with lactose)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
MDL number:
UNSPSC Code:
23151817
Recommended Products
form
lyophilized powder (stabilized with lactose)
extent of labeling
4-6 mg per mL
matrix
cross-linked 4% beaded agarose
capacity
3-5 mg/mL binding capacity (human serum albumin)
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
Quantity
Swelling: 1 g swells to approx. 5 ml
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service