Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

R8144

Sigma-Aldrich

Monoclonal Anti-HLA-DR−Quantum Red antibody produced in mouse

clone HK14, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Monoclonal Anti-HLA-DR

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

mouse

conjugate

Quantum Red® conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

HK14, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

flow cytometry: 10 μL using 1 × 106 cells

isotype

IgG2a

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

The previously assigned protein identifier P01912 has been merged into P01911. Full details can be found on the UniProt database.

Specificity

Recognizes HLA-DR, a product of the human major histocompatibility complex. The antibody is specific for a non-polymorphic HLA-DR determinant.

Immunogen

GM-1500 human tumor cell line

Target description

HLA-DR is located on chromosome 6. It is composed of a heavy and light chain, which are two non-covalently associated subunits of molecular weight 33-36 kDa and 24-30 kDa, respectively. These class II antigens are expressed primarily on B cells, and on monocytes and activated T cells, and function in cell-cell recognition in regulation of the immune response.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 15 mM sodium azide

Preparation Note

Quantum Red is a tandem fluorescent dye created by coupling R-phycoerythrin with Cy5 reactive dye. This red dye is efficiently excited at 488 nm and emits at 670 nm.

Legal Information

Texas Red is a registered trademark of Life Technologies

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Claudio Mauro et al.
Cell metabolism, 25(3), 593-609 (2017-02-14)
Low-grade systemic inflammation associated to obesity leads to cardiovascular complications, caused partly by infiltration of adipose and vascular tissue by effector T cells. The signals leading to T cell differentiation and tissue infiltration during obesity are poorly understood. We tested whether saturated

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service