Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

R7527

Sigma-Aldrich

Monoclonal Anti-Rem (Ges) antibody produced in mouse

clone 24E4, purified immunoglobulin, buffered aqueous glycerol solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

24E4, monoclonal

form

buffered aqueous glycerol solution

mol wt

antigen 37 kDa

species reactivity

mouse, human

technique(s)

immunoprecipitation (IP): suitable
western blot: 1 μg/mL using whole cell extract of C2C12 cells, a mouse muscle myoblast cell line.

isotype

IgG1

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... REM1(28954)
mouse ... Rem1(19700)

Immunogen

recombinant mouse Rem.

Physical form

Solution in 20 mM sodium phosphate, 150 mM sodium chloride, 50% glycerol, pH 7.5, containing 3 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service