Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

R7404

Sigma-Aldrich

Anti-RAD9 antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-hRAD9

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 50 kDa (also band at 60 kDa)

species reactivity

human

technique(s)

western blot: 1:500-1:1,000 using extracts of 293T cells transfected with RAD9

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RAD9A(5883)

General description

RAD9 checkpoint clamp component A (RAD9A), an important member of the 911 complex is mapped to human chromosome 11q13.

Specificity

Anti-RAD9 specifically recognizes recombinant RAD9 (50 and 60 kDa).

Immunogen

synthetic peptide corresponding to amino acid residues 376-391 of human RAD9, conjugated to KLH via an N-terminal added cysteine residue. The immunizing peptide corresponds to RAD9A and is not present in RAD9B.

Application

Anti-RAD9 antibody produced in rabbit may be used in western blotting.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Biochem/physiol Actions

RAD9 checkpoint clamp component A (RAD9A) participates in human cellular responses to DNA damage. RAD9 is found to possess 3′ to 5′ exonuclease activity, which may contribute to its role in sensing and repairing DNA damage. RAD9 can be phosphorylated by ataxia-telangiectasia mutated (ATM) in response to DNA damage. It helps to stimulate S-phase checkpoint by modulating the phosphorylation of cell cycle checkpoint protein (RAD17), checkpoint kinases (Chks 1 and 2) Chk1, Chk2 and structure maintenance of chromosome 1 (Smc1) in response to replication block and DNA damage. It helps to maintain the stability of genome. In addition to its DNA repair role, it also plays a major role in radio resistance, androgen receptor signaling repression and immunoglobulin class switching. RAD9 also participates in telomere maintenance.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing, or storage in “frost-free” freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Human Rad9 is required for the activation of S-phase checkpoint and the maintenance of chromosomal stability
Dang T, et al.
Genes Cells, 10(4), 287-295 (2005)
Rad9a is involved in chromatin decondensation and post-zygotic embryo development in mice
Huang L, et al.
Cell Death and Differentiation, 26(5), 969-980 (2019)
Contributions of Rad9 to tumorigenesis
Broustas CG and Lieberman HB
Journal of Cellular Biochemistry, 113, 742-751 (2012)
The Rad9A checkpoint protein is required for nuclear localization of the claspin adaptor protein
Sierant ML, et al.
Cell Cycle, 9(3), 548-556 (2010)
TOPBP1 recruits TOP2A to ultra-fine anaphase bridges to aid in their resolution
Broderick R, et al.
Nature Communications, 6(1), 1-12 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service