Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

R7154

Sigma-Aldrich

Anti-RAD1, N-terminal antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-Cell Cycle Checkpoint Protein hRad1, Anti-DNA Repair Exonuclease (REC1), Anti-DNA Repair Protein RAD1, Anti-Rad1-like DNA Damage Checkpoint Protein

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 30 kDa

species reactivity

human

technique(s)

immunoprecipitation (IP): 5-10 μL using HeLa cell lysates
western blot: 1:250-1:500 using extracts of HL-60 cells

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RAD1(5810)

General description

RAD1 checkpoint DNA exonuclease/RAD1 is located on human chromosome 5p13.2. It is a member of the Rad family of proteins.

Specificity

Anti-RAD1, N-terminal, specifically recognizes RAD1.

Immunogen

synthetic peptide corresponding to amino acids 2-19 of human RAD1, conjugated to KLH via a C-terminal added cysteine residue. The immunizing peptide is present in RAD1A and RAD1B, isoforms 1 and 2.

Application

Anti-RAD1, C-terminal antibody produced in rabbit may be used in immunoblotting and immunoprecipitation.

Biochem/physiol Actions

RAD1 checkpoint DNA exonuclease/RAD1 can form a stable radioresponsive checkpoint complex commonly known as 9-1-1, which participates in cellular responses to DNA damage or inhibition of DNA synthesis. RadI can serve as a DNA damage sensor. It exhibits terminal exonuclease activity on double-stranded DNA with a preference for 3-prime ends. RAD1 can modulate specifically the expansion of minisatellite tracts.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing, or storage in “frost-free” freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog, our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

RAD1 Controls the Meiotic Expansion of the Human HRAS1 Minisatellite in Saccharomyces cerevisiae
Jauert PA, et al.
Molecular and Cellular Biology, 22(3), 953-964 (2002)
Human hRad1 but not hRad9 protects hHus1 from ubiquitin-proteasomal degradation
Hirai I, et al.
Oncogene, 23(30), 5124-5130 (2004)
U K Marathi et al.
Genomics, 54(2), 344-347 (1998-11-26)
Cell cycle checkpoints are gating mechanisms that govern cell cycle progression in the presence of DNA damage and incomplete DNA replication. The Schizosaccharomyces pombe Rad1 protein is an essential component of cell cycle checkpoints activated by both types of genomic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service