R6018
Ribonucleic acid, transfer, lysine specific from Escherichia coli
lyophilized powder
Synonym(s):
E. coli Lys tRNA, tRNA
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
grade
Molecular Biology
form
lyophilized powder
storage temp.
−20°C
General description
Transfer RNA is isolated from E. coli. This transfer RNA is specific for transfer of the amino acid lysine. The sequence and structure has been published previously.
Application
Suitable for use:
- As a tRNA standard
- As a substrate in tRNA synthatase assays
- As a substrate in other reactions involving tRNAs
related product
Product No.
Description
Pricing
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Mridusmita Saikia et al.
RNA (New York, N.Y.), 16(7), 1317-1327 (2010-05-21)
The N(1)-methyl-Adenosine (m(1)A58) modification at the conserved nucleotide 58 in the TPsiC loop is present in most eukaryotic tRNAs. In yeast, m(1)A58 modification is essential for viability because it is required for the stability of the initiator-tRNA(Met). However, m(1)A58 modification
Tissue-specific differences in human transfer RNA expression.
Dittmar, K.A., et al.
PLoS Genetics, 2, 2107-2107 (2006)
V L Sharygin et al.
Radiatsionnaia biologiia, radioecologiia, 53(2), 151-169 (2013-06-22)
The responses of deoxyribonucleotide (dNTP), DNA and protein synthesis systems in blood-forming organs of animals (dogs, mice) as well as changes in Fe(3+)-transferrin (Fe(3+)-TF) and Cu(2+)-ceruloplasmin (Cu(2+)-CP) pools in blood to gamma-irradiation and the administration of radioprotectors have been studied.
Hanjoo Kim et al.
Nucleic acids research, 41(Web Server issue), W492-W498 (2013-06-14)
To facilitate the analysis of large-scale high-throughput capillary electrophoresis data, we previously proposed a suite of efficient analysis software named HiTRACE (High Throughput Robust Analysis of Capillary Electrophoresis). HiTRACE has been used extensively for quantitating data from RNA and DNA
North, south, or east? Blotting techniques.
M W Nicholas et al.
The Journal of investigative dermatology, 133(7), e10-e10 (2013-06-14)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service