Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

R4905

Sigma-Aldrich

Monoclonal Anti-Rin/Rit2 antibody produced in mouse

~1 mg/mL, clone 18G4(27G2), purified immunoglobulin, buffered aqueous glycerol solution

Synonym(s):

Anti-Ras-like without CAAX 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

18G4(27G2), monoclonal

form

buffered aqueous glycerol solution

mol wt

antigen ~36 kDa

species reactivity

human, mouse

concentration

~1 mg/mL

technique(s)

western blot: 1:5,000 using HEK293 cells expressing HA-tagged Rit

isotype

IgG2b

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RIT2(6014)
mouse ... Rit2(19762)

General description

Rin/Rit2 (Ras-like without CAAX 2) belongs to the Ras superfamily of small guanosine triphosphatases. In human, it is expressed in several neurons whereas in mouse, it is expressed in the ganglion cell layer and inner nuclear layer of the retina. It is located on human chromosome 18q12.3.

Immunogen

recombinant human Rin.

Biochem/physiol Actions

Aberrations in Rin/Rit2 (Ras-like without CAAX 2) causes parkinson′s disease. It participates in neurotrophin signaling. It trigger ERK (extracellular signal-regulated kinase) and p38 MAP (mitogen-activated protein) kinase signaling to promote differentiation.

Physical form

Solution in 20 mM sodium phosphate, 150 mM sodium chloride, 50% glycerol, and 3 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

RIT2 polymorphism is associated with Parkinson's disease in a Han Chinese population
Nie K, et al.
Neurobiology of Aging, 36(3) (2015)
RIT2, a neuron-specific small guanosine triphosphatase, is expressed in retinal neuronal cells and its promoter is modulated by the POU4 transcription factors
Zhang L,et al.
Molecular Vision, 19, 1371-1386 (2013)
Rin GTPase couples nerve growth factor signaling to p38 and b-Raf/ERK pathways to promote neuronal differentiation
Shi GX, et al.
The Journal of Biological Chemistry, 280(45), 37599-37609 (2005)
Ling Zhang et al.
Molecular vision, 19, 1371-1386 (2013-06-28)
Ras-like without CAAX 2 (RIT2), a member of the Ras superfamily of small guanosine triphosphatases, is involved in regulating neuronal function. RIT2 is a unique member of the Ras family in that RIT2 is preferentially expressed in various neurons, including

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service