synthetic peptide corresponding to amino acids 322-340 of rat reelin, conjugated to KLH. This sequence is highly conserved in mouse reelin (single amino acid substitution) and human reelin (84% identity).
Target description
Reelin encodes a large secreted extracellular matrix protein thought to control cell-cell interactions critical for cell positioning and neuronal migration during brain development. This protein may be involved in schizophrenia, autism, bipolar disorder,
Physical form
Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% BSA and 15 mM sodium azide.
Storage and Stability
For continuous use, store at 2-8 °C for up to one month. For extended storage freeze in working aliquots. Repeated freezing and thawing, or storage in "frost-free" freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Journal of sleep research, 28(3), e12721-e12721 (2018-07-03)
Fatigue in multiple sclerosis is a very common and cumbersome symptom, but its aetiology is poorly understood. Proteomics is increasingly implemented in multiple sclerosis research, but has not yet been used to study the neurobiological basis of fatigue in multiple
In the hippocampal CA1, caudal ganglionic eminence (CGE)-derived interneurons are recruited by activation of glutamatergic synapses comprising GluA2-containing calcium-impermeable AMPARs and exert inhibitory regulation of the local microcircuit. However, the role played by AMPARs in maturation of the developing circuit
Neurobiology of disease, 129, 208-216 (2018-07-23)
Traumatic brain injury is a major risk factor for many long-term mental health problems. Although underlying mechanisms likely involve compromised inhibition, little is known about how individual subpopulations of interneurons are affected by neurotrauma. Here we report long-term loss of
The Journal of neuroscience : the official journal of the Society for Neuroscience, 39(14), 2635-2648 (2019-02-02)
The maturation of GABAergic inhibitory circuits is necessary for the onset of the critical period for ocular dominance plasticity (ODP) in the postnatal visual cortex (Hensch, 2005; Espinosa and Stryker, 2012). When it is deficient, the critical period does not
Preterm brain injury, occurring in approximately 30% of infants born <32 weeks gestational age, is associated with an increased risk of neurodevelopmental disorders, such as autism spectrum disorder (ASD) and attention deficit hyperactivity disorder (ADHD). The mechanism of gray matter
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.