Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

R4028

Sigma-Aldrich

Anti-phospho-Retinoblastoma (pSer780) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-phospho-Rb (pSer780)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

target post-translational modification

phosphorylation (pSer780)

Gene Information

human ... RB1(5925)

Immunogen

synthetic phosphopeptide derived from the region of human Rb that contains Ser780.

Physical form

Solution in Dulbecco′s phosphate buffered saline (without Mg2+ and Ca2+), pH 7.3, with 1.0 mg/mL BSA (IgG, protease free) with 50% glycerol, and 0.05% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shan Zhu et al.
Journal of cellular biochemistry, 117(6), 1359-1369 (2015-11-04)
Cellular senescence impedes cancer progression by limiting uncontrolled cell proliferation. To identify new genetic events controlling senescence, we performed a small interfering RNA screening human cancer cells and identified a number of targets potentially involved in senescence of MDA-MB-231 human

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service