Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

R3778

Sigma-Aldrich

Anti-phospho-Retinoblastoma (pSer807) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-phospho-Rb (pSer807)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

western blot: 0.25-0.75 μg/mL using Jurkat cells in high growth phase.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

target post-translational modification

phosphorylation (pSer807)

Gene Information

human ... RB1(5925)

General description

RB1 (RB transcriptional corepressor 1) codes for nuclear phosphoprotein pRB (retinoblastoma protein). It is located on chromosome 13q14. It is a tumor-suppressor gene.

Immunogen

synthetic phosphopeptide derived from the region of human Rb that contains Ser807.

Biochem/physiol Actions

RB1 (RB transcriptional corepressor 1) plays an important role throughout G1/S phase transition. Absence of RB induces prostate cancer metastasis. It plays an important role in modulating the cell cycle. It also regulates cell stemness and differentiation.

Physical form

Solution in phosphate buffer, pH 7.4, containing 1 mg/mL BSA (protease and IgG-free) and 0.05% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Parent-of-Origin Effect Impacts the Phenotype in Low Penetrance Retinoblastoma Families Segregating the c.1981C>T/p.Arg661Trp Mutation of RB1
Eloy P, et al.
PLoS Genetics, 12(2) (2016)
Do MRPS18-2 and RB proteins cooperate to control cell stemness and differentiation, preventing cancer development
Kashuba E and Mushtaq M
Experimental Oncology, 39(1), 12-16 (2017)
RB Loss Promotes Prostate Cancer Metastasis
Thangavel C, et al.
Cancer Research, 77(4), 982-995 (2017)
The Prognostic Value of the Immunohistochemical Expression of Phosphorylated RB and p16 Proteins in Association with Cyclin D1 and the p53 Pathway in a Large Cohort of Patients with Breast Cancer Treated with Taxane-based Adjuvant Chemotherapy
Gavressea T, et al.
Anticancer Research, 37(6), 2947-2957 (2017)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service