Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

R3403

Sigma-Aldrich

Anti-phospho-Retinoblastoma (pSer249/pThr252) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-phospho-Rb (pSer249/pThr252)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

rat, human, mouse

technique(s)

western blot: 0.25-1.0 μg/mL using Jurkat cells in high growth phase.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

target post-translational modification

phosphorylation (pSer249/pThr252)

Gene Information

human ... RB1(5925)
mouse ... Rb1(19645)
rat ... Rb1(24708)

Immunogen

synthetic phosphopeptide derived from the region of human Rb that contains Ser249 and Thr252.

Physical form

Solution in phosphate buffer, pH 7.4, containing 1 mg/mL BSA (protease and IgG-free) and 0.05% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaolan Zhu et al.
International journal of cancer, 135(6), 1286-1296 (2014-02-11)
Multidrug resistance (MDR) remains a major obstacle to effective chemotherapy treatment in ovarian cancer. In our study, paclitaxel-resistant ovarian cancer patients and cell lines had decreased miR-145 levels and expressed high levels of Sp1 and Cdk6. Introducing miR-145 into SKOV3/PTX

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service