Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

R3151

Sigma-Aldrich

Monoclonal Anti-Rhodopsin Kinase 1A antibody produced in mouse

clone G8, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-GRK1a

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$614.00

$614.00


Check Cart for Availability


Select a Size

Change View
100 μG
$614.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.44

$614.00


Check Cart for Availability

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

G8, monoclonal

form

buffered aqueous solution

species reactivity

rat, bovine, chicken, human, mouse

technique(s)

immunoprecipitation (IP): suitable
indirect immunofluorescence: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... GRK1(6011)
mouse ... Grk1(24013)
rat ... Grk1(81760)

General description

GRK1 (rhodopsin kinase) is an enzyme that belongs to serine/threonine protein kinase family and expresses mainly in rod and cone photoreceptors. It has a crucial role in phosphorylation of photoactivated rhodopsin and hence starting steps in its deactivation. Monoclonal anti-rhodopsin kinase 1a antibody can be used in immunoprecipitation. Mouse anti-rhodopsin kinase 1a antibody reacts specifically with an epitope present in the C-terminal region of mouse, bovine, rat, chicken and human GRK1a. The product has shown no cross-reactivity with other G protein coupled kinases together with GRK1b.

Immunogen

derived from human GRK1

Application

Monoclonal anti-rhodopsin kinase 1a antibody can be used in indirect immunofluorescence and western blotting.

Target description

Rhodopsin Kinase 1A encodes a member of the guanine nucleotide-binding protein (G protein)-coupled receptor kinase subfamily of the Ser/Thr protein kinase family. The protein phosphorylates rhodopsin and initiates its deactivation.

Physical form

Supplied in phosphate buffered saline containing 0.05% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

X Zhao et al.
FEBS letters, 454(1-2), 115-121 (1999-07-21)
The G protein-coupled receptor kinases (GRKs) are important enzymes in the desensitization of activated G protein-coupled receptors (GPCR). Seven members of the GRK family have been identified to date. Among these enzymes, GRK1 is involved in phototransduction and is the
X Zhao et al.
The Journal of biological chemistry, 273(9), 5124-5131 (1998-03-28)
The G protein-coupled receptor kinases (GRKs) are critical enzymes in the desensitization of activated G protein-coupled receptors. Six members of the GRK family have been identified to date. Among these enzymes, GRK1 (rhodopsin kinase) is involved in phototransduction and is

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service