Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

R2777

Sigma-Aldrich

Monoclonal Anti-Retinoid X Receptor β antibody produced in mouse

~1 mg/mL, clone 147, purified immunoglobulin, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

147, monoclonal

form

buffered aqueous solution

mol wt

antigen ~60 kDa

species reactivity

mouse

concentration

~1 mg/mL

technique(s)

western blot: 1 μg/mL

isotype

IgG1

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... Rxrb(20182)

Immunogen

synthetic peptide derived from the hinge of the mouse RXRβ.

Physical form

Solution in phosphate buffered saline, pH 7.4, containing 0.08% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service