R2031
Relaxin-3 human
recombinant, expressed in E. coli, >95% (SDS-PAGE), lyophilized powder
Synonym(s):
RLN3, RXN3, Relaxin 3
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
CAS Number:
UNSPSC Code:
12352202
biological source
human
recombinant
expressed in E. coli
assay
>95% (SDS-PAGE)
form
lyophilized powder
mol wt
(Contains two disulfide-linked subunits: A chain ~2.5 kDa B chain ~3.0 kDa)
suitability
endotoxin tested
UniProt accession no.
storage temp.
−20°C
Gene Information
human ... RLN3(117579)
General description
Neuropeptide that is related to the insulin family and located in the hypothalamus
Biochem/physiol Actions
Relaxin 3 participates in neuropeptide signaling processes
Relaxin-3 also has been shown to inhibit insulin secretion from pancreatic islet cells.
Physical form
Lyophilized powder containing BSA
Reconstitution
In Phosphate buffered saline solution containing at least 0.1% serum albumin. The concentration should be ≥10 μg/mL
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Inhibitory Effect of Relaxin-3 on Insulin Secretion in Isolated Pancreas and Insulinoma.
Yamamoto, H.
Journal of AOAC (Association of Official Analytical Chemists) International, 55, 132-137 (2009)
Ying Wu et al.
Nature communications, 6, 7708-7708 (2015-07-22)
Effective annual influenza vaccination requires frequent changes in vaccine composition due to both antigenic shift for different subtype hemagglutinins (HAs) and antigenic drift in a particular HA. Here we present a broadly neutralizing human monoclonal antibody with an unusual binding
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service