R1777
Anti-Rit2 antibody produced in rabbit
~1 mg/mL, fractionated antiserum, buffered aqueous solution
Synonym(s):
Anti-Ras-like protein in neurons
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
biological source
rabbit
Quality Level
conjugate
unconjugated
antibody form
fractionated antiserum
antibody product type
primary antibodies
clone
polyclonal
form
buffered aqueous solution
species reactivity
human
concentration
~1 mg/mL
technique(s)
western blot: suitable
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... RIT2(6014)
Immunogen
recombinant His-tagged human Rit2 (previously designated as Rin) protein.
Target description
Rit2, a member of the Ras superfamily of Ras-related GTPases, may be involved in calcium-mediated signaling within neurons.
Physical form
Supplied in sterile-filtered phosphate buffered saline, pH 7.3, containing 0.08% sodium azide.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Not finding the right product?
Try our Product Selector Tool.
Storage Class
10 - Combustible liquids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service