Key Documents
R1766
Microcentrifuge tube rack, Nalgene®
capacity, 96 0.5-mL tubes (8 × 12)
Synonym(s):
test tube rack, two tier tube rack
About This Item
UNSPSC Code:
41103900
Product R1766 is not currently sold in your country. Contact Technical Service
material
glass-filled acetal
feature
capacity 96 0.5-mL tubes (8 × 12)
packaging
pack of 1 ea
manufacturer/tradename
Nalgene 5973-9305
L × W × H
171 mm × 112 mm × 54 mm
General description
Two-tiered design. Holes in upper and lower tiers offer stable support; wide spacing minimizes interference from overlapping lips. Compatible with most laboratory chemicals, except acids. Autoclavable
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)