Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

R1533

Sigma-Aldrich

Rhodblock 3

≥98% (HPLC), powder

Synonym(s):

2-(5-Chloro-6-methylbenzofuran-3-yl)-acetamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H10ClNO2
CAS Number:
Molecular Weight:
223.66
UNSPSC Code:
12352203

assay

≥98% (HPLC)

form

powder

color

white to off-white

solubility

DMSO: ≥50 mg/mL

storage temp.

room temp

Biochem/physiol Actions

Rhodblock 3 is an inhibitor of Rho Kinase pathway.
Rhodblock 3 is an inhibitor of Rho Kinase pathway. Rhodblock 3 acts upstream of Rok in Drosophila, resulting in the downregulation of Rok and inhibition of myosin phosphorylation. Rhodblock 3 inhibits phospho-MRLC localization as well as furrow localization of the septin Peanut and increased Peanut′s localization on microtubules but did not affect other proteins tested, such as Anillin.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service