The Arabidopsis thaliana Rab GTPase (RabA4b) is a Rab GTPase, that is expressed in various plant organs. In A. thaliana, RabA4b is ubiquitously expressed, mainly located at an inverted cone-shaped area in the apical region in the root hairs. The Arabidopsis genome contains a total of 93 small GTPase genes, including 57 members of the Rab family. Arabidopsis thaliana Rab GTPase RabA4b (AtRabA4b, AtGB3, AtRab11G) is highly similar to AtRab11.
synthetic peptide corresponding to amino acids 201-219 located near the C-terminus of Arabidopsis thaliana AtRabA4b fusion protein.
Application
Anti-AtRabA4 antibody produced in rabbit may be used in immunoblotting.
Biochem/physiol Actions
The Arabidopsis thaliana Rab GTPase (RabA4b) can control membrane trafficking steps involved in the polarized deposition of cell wall components in tip-growing root hair cells. AtRabA4b can label the trans-Golgi network compartment displaying a developmentally regulated polar distribution in growing root hair cells of A. thaliana.
Physical form
Solution in 0.01 M phosphate buffered saline, pH 7.4, and 15 mM sodium azide.
Storage and Stability
For continuous use, store at 2-8 °C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing, or storage in “frost-free” freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Interdependence of endomembrane trafficking and actin dynamics during polarized growth of Arabidopsis pollen tubes
Zhang Y, et al.
Plant Physiology, 152(4), 2200-2210 (2010)
The Arabidopsis Rab GTPase RabA4b localizes to the tips of growing root hair cells
Preuss ML, et al.
Plant Cell, 16(6), 1589-1603 (2004)
Recruitment of PLANT U-BOX13 and the PI4K beta1/beta2 phosphatidylinositol-4 kinases by the small GTPase RabA4B plays important roles during salicylic acid-mediated plant defense signaling in Arabidopsis
Antignani V, et al.
Plant Cell, 27(1), 243-261 (2015)
Questions
Reviews
★★★★★ No rating value
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.