QPREST21503
SILu™PrEST IF2P
SILuPrESTs Powered by Atlas Antibodies, buffered aqueous solution
Synonym(s):
Translation initiation factor IF-2
About This Item
recombinant
expressed in E. coli LysA ArgA BL21(DE3)
assay
>80% (SDS-PAGE)
form
buffered aqueous solution
mol wt
predicted mol wt 28kDa including tags
purified by
immobilized metal affinity chromatography (IMAC)
packaging
pkg of 1nmol × 5 vials
storage condition
avoid repeated freeze/thaw cycles
immunogen sequence
QEMADSLGVRIFSAEIIYHLFDAFTKYRQDYKKQKQEEFKHIAVFPCKIKILPQYIFNSRDPIVMGVTVEAGQVKQGTPMCVPSK
Peptide Atlas accession no.
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... IF2P(9669)
General description
Application
Physical form
Preparation Note
Analysis Note
The SILuPrEST standards are produced using metabolic labeling with heavy isotope labeled (15N, 13C) Lysine and Arginine residues, with greater than 99% isotopic label incorporation.
GSSHHHHHHSSGLVPRGSHMASLAEAKVLANRELDKYGVSDYHKNLINNAKTVEGVKDLQAQVVESAKKARISEATDGLSDFLKSQTPAEDTVKSIELAEAKVLANRELDKYGVSDYYKNLINNAKTVEGVKALIDEILAALPGTFAHYMDPNSSSVDKLAAA.
The specific human protein sequence begins directly after ′VDKLAAA′.
Legal Information
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service