Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PZ0401

Sigma-Aldrich

PF-06855800 variable hydrate

≥98% (HPLC)

Synonym(s):

(2R,3R,4S,5R)-2-(4-Amino-7H-pyrrolo[2,3-d]pyrimidin-7-yl)-5-((R)-(4-chloro-3-fluorophenyl)(hydroxy)methyl)tetrahydrofuran-3,4-diol, 7-[(5R)-5-C-(4-Chloro-3-fluorophenyl)-ß-D-ribofuranosyl]-7H-pyrrolo[2,3-d]pyrimidin-4-amine

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$72.50
25 MG
$345.00

$72.50

List Price$85.00Save 15%
Web-Only Promotion

Check Cart for Availability

Request a Bulk Order
Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
5 MG
$72.50
25 MG
$345.00

About This Item

Empirical Formula (Hill Notation):
C17H16ClFN4O4 · xH2O
Molecular Weight:
394.78 (anhydrous basis)
UNSPSC Code:
12352200
NACRES:
NA.77

$72.50

List Price$85.00Save 15%
Web-Only Promotion

Check Cart for Availability

Request a Bulk Order
Notify Me

Get notified when this item is ready to ship via email.

Quality Level

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

PF-06855800 is a cell penetrant, potent and selective S-adenosylmethionine (SAM) competitive inhibitor of the protein arginine methyltransferase 5 (PRMT5) that exhibits anticancer activity in both in vitro and in vivo models.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service