Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PZ0353

Sigma-Aldrich

UK-59811 hydrochloride

≥98% (HPLC)

Synonym(s):

4-(2-Bromophenyl)-2-[[2-(dimethylamino)ethoxy]methyl]-1,4-dihydro-6-methyl-3,5-pyridinedicarboxylic acid 3-ethyl 5-methyl ester hydrochloride, UK59811 hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$49.73
25 MG
$236.00

$49.73

List Price$58.30Save 15%
Web-Only Promotion

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$49.73
25 MG
$236.00

About This Item

Empirical Formula (Hill Notation):
C22H29BrN2O5 · HCl
CAS Number:
Molecular Weight:
517.84
UNSPSC Code:
51111800
NACRES:
NA.77

$49.73

List Price$58.30Save 15%
Web-Only Promotion

Check Cart for Availability

Request a Bulk Order

Quality Level

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

CCOC(C1=C(COCCN(C)C)NC(C)=C(C(OC)=O)C1C2=C(Br)C=CC=C2)=O

InChI

1S/C22H29BrN2O5/c1-6-30-22(27)20-17(13-29-12-11-25(3)4)24-14(2)18(21(26)28-5)19(20)15-9-7-8-10-16(15)23/h7-10,19,24H,6,11-13H2,1-5H3

InChI key

XCBGSNNUPRHJGR-UHFFFAOYSA-N

Related Categories

Biochem/physiol Actions

UK-59811 is a 1,4-dihydropyridine calcium channel blocker. It has been used in a crystallographic and functional analysis of drug binding to the bacterial homotetrameric model voltage-gated calcium channel CaVAb, comparing 1,4-dihydropyridine binding to phenylalkylamine binding. UK-59811 inhibited CaVAb with an IC50 value of 194 nM, binding at the outer, lipid-facing surface of the pore at the interface between two CaVAb subunits, while at concentrations above 1 μM, it also bound in the pore. It was contrasted with phenylalkylamine Br-verapamil which binds on the intracellular side of the selectivity filter in the central cavity of the pore, physically blocking the pore.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lin Tang et al.
Nature, 537(7618), 117-121 (2016-08-25)
Ca2+ antagonist drugs are widely used in therapy of cardiovascular disorders. Three chemical classes of drugs bind to three separate, but allosterically interacting, receptor sites on CaV1.2 channels, the most prominent voltage-gated Ca2+ (CaV) channel type in myocytes in cardiac

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service