Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PZ0350

Sigma-Aldrich

PF-06748962

≥98% (HPLC)

Synonym(s):

3-[(3R)-3-Methyl-2-oxo-3-piperidinyl]-6-(5-methyl-3-quinolinyl)-2(1H)-pyridinone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H21N3O2
CAS Number:
Molecular Weight:
347.41
UNSPSC Code:
12352202
NACRES:
NA.77

Quality Level

assay

≥98% (HPLC)

form

powder

optical activity

[α]/D -880 to -920°, c = 0.5 in chloroform-d

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

room temp

SMILES string

CC1=CC=CC2=NC=C(C3=CC=C([C@]4(C)C(NCCC4)=O)C(N3)=O)C=C21

Biochem/physiol Actions

PF-06748962 is a potent and selective lactam-based prostaglandin EP3 receptor antagonist.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service