Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PZ0293

Sigma-Aldrich

GRN-529

≥97% (HPLC)

Synonym(s):

[4-(Difluoromethoxy)-3-[2-(2-pyridinyl)ethynyl]phenyl](5,7-dihydro-6H-pyrrolo[3,4-b]pyridin-6-yl)-methanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H15F2N3O2
CAS Number:
Molecular Weight:
391.37
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

assay

≥97% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

room temp

SMILES string

FC(F)OC1=C(C#CC2=NC=CC=C2)C=C(C(N3CC(N=CC=C4)=C4C3)=O)C=C1

InChI

1S/C22H15F2N3O2/c23-22(24)29-20-9-7-16(12-15(20)6-8-18-5-1-2-10-25-18)21(28)27-13-17-4-3-11-26-19(17)14-27/h1-5,7,9-12,22H,13-14H2

InChI key

JITMSIRHBAVREW-UHFFFAOYSA-N

Biochem/physiol Actions

GRN-529 is a brain penetrant, potent and selective negative allosteric modulator of the mGluR5 receptor (metabotropic glutamate receptor subtype 5) with an IC50 of 3.1 nM and >1000-fold selectivity vs mGluR1. GRN-529 was shown to alleviate multiple diagnostic behavioral symptoms of autism in BTBR and C58 mice models.

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service