Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PZ0279

Sigma-Aldrich

Sunepitron hydrochloride

Synonym(s):

1-[[(7S,9aS)-Octahydro-2-(2-pyrimidinyl)-2H-pyrido[1,2-a]pyrazin-7-yl]methyl]-2,5-pyrrolidinedione hydrochloride, 1-{[2-(Pyrimidin-2-yl)octahydro-2H-pyrido[1,2-a]pyrazin-7-yl]methyl}pyrrolidine-2,5-dione, CP 93393 hydrochloride, CP-93,393

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H23N5O2 · HCl
CAS Number:
Molecular Weight:
365.86
UNSPSC Code:
12352200
NACRES:
NA.77

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

H2O: 20 mg/mL, clear

storage temp.

room temp

InChI

1S/C17H23N5O2.ClH/c23-15-4-5-16(24)22(15)11-13-2-3-14-12-21(9-8-20(14)10-13)17-18-6-1-7-19-17;/h1,6-7,13-14H,2-5,8-12H2;1H/t13-,14-;/m0./s1

InChI key

HWGPYTRJVVMHGT-IODNYQNNSA-N

Biochem/physiol Actions

Sunepitron hydrochloride is a combined serotonin 5-HT1A receptor agonist and α2-adrenergic receptor antagonist. Sunepitron hydrochloride has been tested for the treatment of depression and anxiety.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service