Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PZ0232

Sigma-Aldrich

PF-04471141 hydrochloride

≥98% (HPLC)

Synonym(s):

4-(3-Pentylamino)-6,7,8-trimethoxyquinazoline hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$123.00
25 MG
$512.00

$123.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$123.00
25 MG
$512.00

About This Item

Empirical Formula (Hill Notation):
C16H23N3O3 · xHCl
CAS Number:
Molecular Weight:
305.37 (free base basis)
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.77

$123.00


Check Cart for Availability

Request a Bulk Order

Quality Level

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

−20°C

SMILES string

CCC(CC)NC1=NC=NC2=C(OC)C(OC)=C(OC)C=C21.Cl

InChI

1S/C16H23N3O3.ClH/c1-6-10(7-2)19-16-11-8-12(20-3)14(21-4)15(22-5)13(11)17-9-18-16;/h8-10H,6-7H2,1-5H3,(H,17,18,19);1H

InChI key

QRHFQSUUJMOIHO-UHFFFAOYSA-N

Biochem/physiol Actions

PF-04471141 is a potent, aelective inhibitor of the calcium-activated phosphodiesterase PDE1.
PF-04471141 is a potent, selective inhibitor of the calcium-activated phosphodiesterase PDE1. (PDE1B IC50 = 35 nM).

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service