Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PZ0226

Sigma-Aldrich

CP 524515

≥98% (HPLC)

Synonym(s):

(2R,4S)-4-[[[3,5-Bis(trifluoromethyl)phenyl]methyl](methoxycarbonyl)amino]-2-ethyl-3,4-dihydro-6-(trifluoromethyl)-1(2H)-quinolinecarboxylic acid methylethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H27F9N2O4
CAS Number:
Molecular Weight:
614.50
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(OC(C)C)N1C2=CC=C(C(F)(F)F)C=C2[C@@H](N(CC3=CC(C(F)(F)F)=CC(C(F)(F)F)=C3)C(OC)=O)C[C@H]1CC

InChI

1S/C27H27F9N2O4/c1-5-19-12-22(20-11-16(25(28,29)30)6-7-21(20)38(19)24(40)42-14(2)3)37(23(39)41-4)13-15-8-17(26(31,32)33)10-18(9-15)27(34,35)36/h6-11,14,19,22H,5,12-13H2,1-4H3/t19-,22+/m1/s1

InChI key

HFFFFTQVPXWDLI-KNQAVFIVSA-N

Biochem/physiol Actions

CP-524515 is a potent inhbitor of cholesteryl ester transfer protein (CETP) that leads to elevated high-density lipoprotein cholesterol levels. CP-532623 is structurally related to Torcetrapib (Cat. No. PZ0170).
CP-524515 is a potent inhbitor of cholesteryl ester transfer protein (CETP).

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yingxue Cui et al.
Acupuncture in medicine : journal of the British Medical Acupuncture Society, 37(4), 237-243 (2019-05-30)
To investigate the anti-atherogenic effect of moxibustion and whether it is mediated through the reverse cholesterol transport process. 8-week-old male apolipoprotein E deficient (ApoE-/- knockout) mice were randomly divided into two groups (n=10 per group): atherosclerosis (AS) and AS plus

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service